siRNA Details
| Gene Symbol: Cep70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023873.3 | NP_076362.2 | 11 | 1052 | ||
| GenBank mRNA | AF242319.1 | - | 11 | 1088 | ||
| AK021095.1 | - | 10 | 1237 | |||
| AK133048.1 | - | 11 | 1058 | |||
| BC050819.1 | - | 11 | 1044 | 6517145 | ||
| BC071260.1 | - | 11 | 1052 | 6336351 | ||

Cep70
Genomic Map