siRNA Details
| Gene Symbol: Arl13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026577.3 | NP_080853.3 | 2 | 414 | ||
| GenBank mRNA | AK016794.1 | - | 2 | 360 | ||
| AK020859.1 | - | 2 | 355 | |||
| AK021270.1 | - | 2 | 415 | |||
| AK051159.1 | - | 2 | 372 | |||
| BC082574.1 | - | 2 | 414 | 30550373 | ||

Arl13b
Genomic Map