siRNA Details
| Gene Symbol: Arl13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026577.3 | NP_080853.3 | 6 | 1036 | ||
| GenBank mRNA | AK016794.1 | - | 6 | 982 | ||
| AK021270.1 | - | 6 | 1038 | |||
| BC082574.1 | - | 6 | 1036 | 30550373 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arl13b| Gene Symbol: Arl13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026577.3 | NP_080853.3 | 6 | 1036 | ||
| GenBank mRNA | AK016794.1 | - | 6 | 982 | ||
| AK021270.1 | - | 6 | 1038 | |||
| BC082574.1 | - | 6 | 1036 | 30550373 | ||