siRNA Details
| Gene Symbol: 1700067P10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026625.2 | NP_080901.2 | 3 | 323 | ||
| GenBank mRNA | AK015495.1 | - | 3 | 285 | ||
| AK018887.1 | - | 3 | 312 | |||
| BC048440.1 | - | 3 | 313 | 6741836 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700067P10Rik| Gene Symbol: 1700067P10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026625.2 | NP_080901.2 | 3 | 323 | ||
| GenBank mRNA | AK015495.1 | - | 3 | 285 | ||
| AK018887.1 | - | 3 | 312 | |||
| BC048440.1 | - | 3 | 313 | 6741836 | ||