siRNA Details
| Gene Symbol: Tmem86b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023440.2 | NP_075929.1 | 3 | 603 | ||
| GenBank mRNA | BC100527.1 | - | 3 | 603 | 30920275 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem86b| Gene Symbol: Tmem86b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023440.2 | NP_075929.1 | 3 | 603 | ||
| GenBank mRNA | BC100527.1 | - | 3 | 603 | 30920275 | |