siRNA Details
| Gene Symbol: Ift80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026641.2 | NP_080917.1 | 12 | 1370 | ||
| GenBank mRNA | AK042231.1 | - | 12 | 1295 | ||
| AK129339.1 | - | 12 | 1298 | |||
| AK147305.1 | - | 11 | 4897 | |||
| BC013814.1 | - | 7 | 659 | |||
| BC034101.1 | - | 12 | 1265 | 4501451 | ||

Ift80
Genomic Map