siRNA Details
| Gene Symbol: Rbm28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133925.2 | NP_598686.2 | 16 | 1841 | ||
| GenBank mRNA | AK156773.1 | - | 16 | 1778 | ||
| AK161670.1 | - | 16 | 1808 | |||
| AK167050.1 | - | 16 | 1843 | |||
| BC018373.1 | - | 12 | 1215 | 3708613 | ||
| BC023293.1 | - | 6 | 590 | |||
| BC024602.1 | - | 16 | 1722 | |||
| BC027123.1 | - | 9 | 1147 | 5356457 | ||
| BC040811.1 | - | 16 | 1786 | 4015830 | ||

Rbm28
Genomic Map