siRNA Details
| Gene Symbol: Rbm28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133925.2 | NP_598686.2 | 19 | 2345 | ||
| GenBank mRNA | AK156773.1 | - | 19 | 2282 | ||
| AK161670.1 | - | 19 | 2315 | |||
| AK167050.1 | - | 19 | 2347 | |||
| BC018373.1 | - | 15 | 1719 | 3708613 | ||
| BC023293.1 | - | 9 | 1094 | |||
| BC024602.1 | - | 19 | 2226 | |||
| BC027123.1 | - | 12 | 1651 | 5356457 | ||
| BC040811.1 | - | 19 | 2290 | 4015830 | ||

Rbm28
Genomic Map