siRNA Details
| Gene Symbol: Vps53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026664.3 | NP_080940.2 | 4 | 380 | ||
| GenBank mRNA | AK033495.1 | - | 4 | 381 | ||
| AK049259.1 | - | 4 | 199 | |||
| AK149966.1 | - | 4 | 378 | |||
| AK150001.1 | - | 4 | 378 | |||
| AK159454.1 | - | 4 | 359 | |||
| AK166234.1 | - | 4 | 343 | |||
| AK167310.1 | - | 4 | 381 | |||
| BC034371.1 | - | 4 | 374 | 3964008 | ||

Vps53
Genomic Map