siRNA Details
Gene Symbol: Vps53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026664.3 | NP_080940.2 | 8 | 786 | ||
GenBank mRNA | AK019408.1 | - | 8 | 787 | ||
AK033495.1 | - | 8 | 787 | |||
AK049259.1 | - | 8 | 605 | |||
AK149966.1 | - | 8 | 784 | |||
AK150001.1 | - | 8 | 784 | |||
AK159454.1 | - | 8 | 765 | |||
AK167310.1 | - | 8 | 787 |
Gene Symbol: Glod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK138073.1 | - | 910 |
