siRNA Details
| Gene Symbol: Kdelc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212445.2 | NP_997610.1 | 8 | 1496 | ||
| GenBank mRNA | AK019513.1 | - | 3 | 244 | ||
| AK075811.1 | - | 8 | 1305 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kdelc2| Gene Symbol: Kdelc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212445.2 | NP_997610.1 | 8 | 1496 | ||
| GenBank mRNA | AK019513.1 | - | 3 | 244 | ||
| AK075811.1 | - | 8 | 1305 | |||