siRNA Details
| Gene Symbol: Nudt22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026675.2 | NP_080951.1 | 3 | 578 | ||
| GenBank mRNA | AK002252.1 | - | 3 | 579 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nudt22| Gene Symbol: Nudt22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026675.2 | NP_080951.1 | 3 | 578 | ||
| GenBank mRNA | AK002252.1 | - | 3 | 579 | ||