siRNA Details
| Gene Symbol: Tlcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291235.1 | NP_001278164.1 | 4 | 270 | ||
| NM_001291236.1 | NP_001278165.1 | 4 | 746 | |||
| NM_001291237.1 | NP_001278166.1 | 5 | 454 | |||
| NM_026708.2 | NP_080984.1 | 4 | 581 | |||
| GenBank mRNA | AK002265.1 | - | 4 | 529 | ||
| AK046577.1 | - | 5 | 2461 | |||
| AK161721.1 | - | 481 | ||||
| AK165432.1 | - | 693 | ||||
| BC005702.1 | - | 4 | 473 | 3598444 | ||
| BC039182.1 | - | 4 | 716 | 3500908 | ||
| BG083251.2 | - | 4 | 328 | |||

Tlcd1
Genomic Map