siRNA Details
Gene Symbol: Slc39a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290765.1 | NP_001277694.1 | 5, 6 | 842 | ||
NM_026721.3 | NP_080997.1 | 5, 6 | 803 | |||
GenBank mRNA | AK035296.1 | - | 5, 6 | 710 | ||
AK156951.1 | - | 5, 6 | 728 | |||
BC100463.1 | - | 5, 6 | 810 | 30918840 | ||
BC125525.1 | - | 5, 6 | 664 | 40130040 | ||
BC137752.1 | - | 5, 6 | 664 | 8860771 |
