siRNA Details
| Gene Symbol: Slc39a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290765.1 | NP_001277694.1 | 8 | 1036 | ||
| NM_026721.3 | NP_080997.1 | 8 | 997 | |||
| GenBank mRNA | AK003191.1 | - | 4 | 299 | ||
| AK035296.1 | - | 8 | 904 | |||
| BC100463.1 | - | 8 | 1004 | 30918840 | ||

Slc39a13
Genomic Map