siRNA Details
| Gene Symbol: Dhrs7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013013.2 | NP_001013031.2 | 6 | 735 | ||
| GenBank mRNA | AI391290.1 | - | 3 | 224 | ||
| BC039565.1 | - | 6 | 626 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhrs7c| Gene Symbol: Dhrs7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013013.2 | NP_001013031.2 | 6 | 735 | ||
| GenBank mRNA | AI391290.1 | - | 3 | 224 | ||
| BC039565.1 | - | 6 | 626 | |||