siRNA Details
| Gene Symbol: Evc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145920.3 | NP_666032.1 | 16 | 2987 | ||
| GenBank mRNA | AB083066.1 | - | 16 | 2917 | ||
| AK003753.1 | - | 3 | 489 | |||
| BC037473.1 | - | 16 | 2968 | 4237472 | ||
| BC064473.1 | - | 16 | 2978 | 3156380 | ||

Evc2
Genomic Map