siRNA Details
| Gene Symbol: Evc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145920.3 | NP_666032.1 | 11 | 1736 | ||
| GenBank mRNA | AB083066.1 | - | 11 | 1666 | ||
| AK044977.1 | - | 11 | 1607 | |||
| AK134298.1 | - | 11 | 1719 | |||
| AK165051.1 | - | 11 | 1621 | |||
| BC037473.1 | - | 11 | 1717 | 4237472 | ||
| BC064473.1 | - | 11 | 1727 | 3156380 | ||

Evc2
Genomic Map