siRNA Details
| Gene Symbol: Gpr155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190297.2 | NP_001177226.1 | 3 | 801 | ||
| NM_001276443.1 | NP_001263372.1 | 3 | 801 | |||
| NM_001276444.1 | NP_001263373.1 | 3 | 801 | |||
| GenBank mRNA | AK089436.1 | - | 3 | 719 | ||
| BC115698.1 | - | 3 | 697 | 40060105 | ||
| HM161763.2 | - | 3 | 778 | |||
| HM161764.2 | - | 3 | 778 | |||
| HM161765.2 | - | 3 | 778 | |||
| HM161766.2 | - | 3 | 778 | |||
| KF358283.1 | - | 3 | 778 | |||
| KF358284.1 | - | 3 | 778 | |||

Gpr155
Genomic Map