siRNA Details
| Gene Symbol: Tmem109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134142.1 | NP_598903.1 | 2 | 202 | ||
| GenBank mRNA | AK049148.1 | - | 2 | 206 | ||
| AK150779.1 | - | 2 | 186 | |||
| AK151850.1 | - | 2 | 186 | |||
| AK152419.1 | - | 2 | 186 | |||
| AK165990.1 | - | 2 | 160 | |||
| BC022997.1 | - | 2 | 202 | 5362560 | ||
| BC023901.1 | - | 2 | 160 | 5321274 | ||
| BC025033.1 | - | 2 | 172 | 4947460 | ||
| BC025815.1 | - | 2 | 194 | 5097938 | ||

Tmem109
Genomic Map