siRNA Details
| Gene Symbol: Col6a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026763.2 | NP_081039.2 | 33 | 5655 | ||
| GenBank mRNA | AB370265.1 | - | 32 | 5747 | ||
| AM231153.1 | - | 27 | 3035 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col6a4| Gene Symbol: Col6a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026763.2 | NP_081039.2 | 33 | 5655 | ||
| GenBank mRNA | AB370265.1 | - | 32 | 5747 | ||
| AM231153.1 | - | 27 | 3035 | |||