siRNA Details
| Gene Symbol: Uckl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026765.3 | NP_081041.2 | 2, 3 | 299 | ||
| GenBank mRNA | AK144803.1 | - | 2, 3 | 305 | ||
| BC016535.1 | - | 2, 3 | 299 | 2655873 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uckl1| Gene Symbol: Uckl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026765.3 | NP_081041.2 | 2, 3 | 299 | ||
| GenBank mRNA | AK144803.1 | - | 2, 3 | 305 | ||
| BC016535.1 | - | 2, 3 | 299 | 2655873 | ||