siRNA Details
| Gene Symbol: Ankra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271388.1 | NP_001258317.1 | 6 | 1206 | ||
| NM_001271389.1 | NP_001258318.1 | 6 | 1206 | |||
| NM_001271390.1 | NP_001258319.1 | 5 | 824 | |||
| NM_001271391.1 | NP_001258320.1 | 5 | 824 | |||
| NM_001271392.1 | NP_001258321.1 | 5 | 824 | |||
| NM_023472.2 | NP_075961.1 | 6 | 1206 | |||
| GenBank mRNA | AB221648.1 | - | 5 | 709 | ||
| AF314031.1 | - | 6 | 1040 | |||
| AK003438.2 | - | 5 | 743 | |||
| AK054037.1 | - | 6 | 1012 | |||
| AK081695.1 | - | 5 | 709 | |||
| AK139343.1 | - | 6 | 1088 | |||
| AK161446.1 | - | 6 | 1212 | |||
| BC037437.1 | - | 5 | 752 | |||
| BC066113.1 | - | 5 | 743 | 30361246 | ||
| BC079615.1 | - | 6 | 1023 | 30361702 | ||

Ankra2
Genomic Map