siRNA Details
| Gene Symbol: Pdrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178939.2 | NP_849270.1 | 1, 2 | 156 | ||
| GenBank mRNA | AK003410.1 | - | 1, 2 | 158 | ||
| AK145939.1 | - | 1, 2 | 201 | |||
| AY286302.1 | - | 1, 2 | 137 | |||
| BC031699.1 | - | 1, 2 | 156 | 4020218 | ||

Pdrg1
Genomic Map