siRNA Details
| Gene Symbol: Ppm1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176833.4 | NP_789803.1 | 6, 7 | 1107 | ||
| GenBank mRNA | AK016942.1 | - | 6, 7 | 1112 | ||
| AK154192.1 | - | 6, 7 | 1010 | |||
| AK164964.1 | - | 6, 7 | 988 | |||
| BC042570.1 | - | 5, 6 | 956 | 4241448 | ||

Ppm1f
Genomic Map