siRNA Details
| Gene Symbol: Ppm1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176833.4 | NP_789803.1 | 6 | 1058 | ||
| GenBank mRNA | AK016942.1 | - | 6 | 1063 | ||
| AK154192.1 | - | 6 | 961 | |||
| AK164964.1 | - | 6 | 939 | |||
| BC042570.1 | - | 5 | 907 | 4241448 | ||

Ppm1f
Genomic Map