siRNA Details
Gene Symbol: Letmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134093.2 | NP_598854.1 | 4, 5 | 568 | ||
GenBank mRNA | AF287293.1 | - | 4, 5 | 508 | ||
AF401483.1 | - | 4, 5 | 401 | |||
AK041312.1 | - | 3, 4 | 897 | |||
AK045256.1 | - | 4, 5 | 388 | |||
AK046496.1 | - | 3, 4 | 372 | |||
AK080221.1 | - | 4, 5 | 488 | |||
AK154676.1 | - | 4, 5 | 483 | |||
AK158718.1 | - | 3, 4 | 345 | |||
AK169341.1 | - | 4, 5 | 488 | |||
AK185638.1 | - | 2, 3 | 112 | |||
AK217327.1 | - | 2, 3 | 112 | |||
BC021361.1 | - | 1, 2 | 240 | 5010892 |
