siRNA Details
| Gene Symbol: Letmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134093.2 | NP_598854.1 | 2 | 350 | ||
| GenBank mRNA | AF287293.1 | - | 2 | 290 | ||
| AF401483.1 | - | 2 | 183 | |||
| AK032864.1 | - | 1 | 707 | |||
| AK041312.1 | - | 1 | 679 | |||
| AK045256.1 | - | 2 | 170 | |||
| AK046496.1 | - | 2 | 270 | |||
| AK080221.1 | - | 2 | 270 | |||
| AK154676.1 | - | 2 | 265 | |||
| AK158718.1 | - | 2 | 243 | |||
| AK169341.1 | - | 2 | 270 | |||
| BY293457.1 | - | 2 | 350 | |||

Letmd1
Genomic Map