siRNA Details
| Gene Symbol: Tm2d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026795.3 | NP_081071.1 | 4 | 529 | ||
| NM_178056.3 | NP_835157.1 | 5 | 622 | |||
| GenBank mRNA | AK003917.1 | - | 4 | 488 | ||
| AK077858.1 | - | 1236 | ||||
| BC024620.1 | - | 4 | 482 | 4192712 | ||

Tm2d3
Genomic Map