siRNA Details
| Gene Symbol: Fam198b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133187.3 | NP_573450.2 | 3 | 1562 | ||
| GenBank mRNA | AF285091.1 | - | 3 | 1504 | ||
| AK052367.1 | - | 3 | 529 | |||
| AK143522.1 | - | 3 | 1293 | |||
| AK195202.1 | - | 1 | 40 | |||
| BC056975.1 | - | 3 | 1331 | 6827227 | ||
| EU797522.1 | - | 3 | 1340 | |||

Fam198b
Genomic Map