siRNA Details
Gene Symbol: Fam198b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133187.3 | NP_573450.2 | 2 | 1392 | ||
GenBank mRNA | AF285091.1 | - | 2 | 1334 | ||
AK052367.1 | - | 2 | 359 | |||
AK143522.1 | - | 2 | 1123 | |||
BC056975.1 | - | 2 | 1161 | 6827227 | ||
EU797522.1 | - | 2 | 1170 |
