siRNA Details
| Gene Symbol: Svop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026805.1 | NP_081081.1 | 10, 11 | 1174 | ||
| GenBank mRNA | AK047655.1 | - | 10, 11 | 1167 | ||
| AK047764.1 | - | 10, 11 | 1175 | |||
| AK053833.1 | - | 10, 11 | 1165 | |||
| AK082744.1 | - | 10, 11 | 1176 | |||
| BC059878.1 | - | 10, 11 | 1127 | 6812082 | ||

Svop
Genomic Map