siRNA Details
Gene Symbol: Tmem219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026827.1 | NP_081103.1 | 3 | 353 | ||
NM_028389.1 | NP_082665.1 | 3 | 350 | |||
GenBank mRNA | AK004043.1 | - | 2 | 313 | ||
AK013646.1 | - | 2 | 155 | |||
AK152526.1 | - | 3 | 315 | |||
BC023442.1 | - | 3 | 328 | 5026775 | ||
BC046763.1 | - | 2 | 495 | 5693779 |
