siRNA Details
| Gene Symbol: Abhd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190437.1 | NP_001177366.1 | 5 | 708 | ||
| NM_145215.2 | NP_660250.1 | 5 | 1185 | |||
| GenBank mRNA | AF412033.1 | - | 5 | 1185 | ||
| AK004244.1 | - | 5 | 719 | |||
| BC016241.1 | - | 3 | 759 | |||
| BC038052.1 | - | 4 | 587 | |||
| BC051486.1 | - | 2 | 258 | |||
| BC056610.1 | - | 5 | 847 | |||
| BC056624.1 | - | 4 | 1170 | |||
| BC069865.1 | - | 2 | 1172 | |||
| BC069866.1 | - | 5 | 807 | 6478980 | ||

Abhd11
Genomic Map