siRNA Details
| Gene Symbol: Abhd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190437.1 | NP_001177366.1 | 1, 2 | 75 | ||
| NM_145215.2 | NP_660250.1 | 1, 2 | 552 | |||
| GenBank mRNA | AF412033.1 | - | 1, 2 | 552 | ||
| AK004244.1 | - | 1, 2 | 87 | |||
| AK041432.1 | - | 114 | ||||
| AK041742.1 | - | 114 | ||||
| BC051486.1 | - | 5 | 891 | |||
| BC056610.1 | - | 2 | 245 | |||
| BC056624.1 | - | 1 | 537 | |||
| BC069865.1 | - | 1 | 287 | |||
| BC069866.1 | - | 1, 2 | 174 | 6478980 | ||

Abhd11
Genomic Map