siRNA Details
| Gene Symbol: Abhd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190437.1 | NP_001177366.1 | 4 | 525 | ||
| NM_145215.2 | NP_660250.1 | 4 | 1002 | |||
| GenBank mRNA | AF412033.1 | - | 4 | 1002 | ||
| AK004244.1 | - | 4 | 536 | |||
| BC016241.1 | - | 2 | 576 | |||
| BC038052.1 | - | 3 | 404 | |||
| BC051486.1 | - | 3 | 441 | |||
| BC056610.1 | - | 4 | 664 | |||
| BC056624.1 | - | 3 | 987 | |||
| BC069865.1 | - | 2 | 886 | |||
| BC069866.1 | - | 4 | 624 | 6478980 | ||

Abhd11
Genomic Map