siRNA Details
| Gene Symbol: Cdhr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024478.1 | NP_001019649.1 | 12 | 1622 | ||
| GenBank mRNA | AK047666.1 | - | 12 | 1623 | ||
| AK049806.1 | - | 12 | 1623 | |||
| BC138911.1 | - | 12 | 1550 | 8861933 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdhr3| Gene Symbol: Cdhr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024478.1 | NP_001019649.1 | 12 | 1622 | ||
| GenBank mRNA | AK047666.1 | - | 12 | 1623 | ||
| AK049806.1 | - | 12 | 1623 | |||
| BC138911.1 | - | 12 | 1550 | 8861933 | ||