siRNA Details
| Gene Symbol: Phtf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172992.3 | NP_766580.3 | 7 | 753 | ||
| GenBank mRNA | AK035536.1 | - | 7 | 707 | ||
| AK037021.1 | - | 7 | 640 | |||
| AK042382.1 | - | 7 | 732 | |||
| AK152951.1 | - | 7 | 706 | |||
| AK163376.1 | - | 7 | 679 | |||
| BC052841.1 | - | 7 | 666 | 30050876 | ||

Phtf2
Genomic Map