siRNA Details
| Gene Symbol: Atp6v1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159632.1 | NP_001153104.1 | 11 | 977 | ||
| NM_133699.2 | NP_598460.1 | 10 | 947 | |||
| GenBank mRNA | AB088357.1 | - | 10 | 932 | ||
| AK004157.1 | - | 7 | 662 | |||
| BC056636.1 | - | 11 | 950 | 5065583 | ||
| BC064071.1 | - | 10 | 916 | |||

Atp6v1c2
Genomic Map