siRNA Details
| Gene Symbol: Atp6v1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159632.1 | NP_001153104.1 | 12 | 1045 | ||
| NM_133699.2 | NP_598460.1 | 11 | 1015 | |||
| GenBank mRNA | AB088357.1 | - | 11 | 1000 | ||
| AK004157.1 | - | 8 | 730 | |||
| BC056636.1 | - | 12 | 1018 | 5065583 | ||

Atp6v1c2
Genomic Map