siRNA Details
Gene Symbol: Tmem214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144525.3 | NP_653108.2 | 8 | 952 | ||
GenBank mRNA | AK034883.1 | - | 8 | 954 | ||
AK053521.1 | - | 1242 | ||||
AK143078.1 | - | 954 | ||||
AK165285.1 | - | 7 | 1115 | |||
AK166510.1 | - | 8 | 947 | |||
AK190374.1 | - | 2 | 170 | |||
BC022142.1 | - | 4 | 229 | 5101205 | ||
BC026651.1 | - | 8 | 868 | 4987771 | ||
BC027046.1 | - | 2 | 246 | 5362953 | ||
BC029150.1 | - | 8 | 940 | 4954376 |
