siRNA Details
Gene Symbol: Tmem214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144525.3 | NP_653108.2 | 17 | 2037 | ||
GenBank mRNA | AK034883.1 | - | 17 | 2039 | ||
AK053521.1 | - | 2327 | ||||
AK165285.1 | - | 15 | 3021 | |||
AK166510.1 | - | 17 | 2032 | |||
BC022142.1 | - | 13 | 1314 | 5101205 | ||
BC022602.1 | - | 11 | 1183 | 4161204 | ||
BC026651.1 | - | 17 | 1953 | 4987771 | ||
BC027046.1 | - | 11 | 1331 | 5362953 | ||
BC029150.1 | - | 17 | 2025 | 4954376 |
