siRNA Details
| Gene Symbol: Tmem214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144525.3 | NP_653108.2 | 13 | 1482 | ||
| GenBank mRNA | AK034883.1 | - | 13 | 1484 | ||
| AK053521.1 | - | 1772 | ||||
| AK143078.1 | - | 1484 | ||||
| AK165285.1 | - | 12 | 1645 | |||
| AK166510.1 | - | 13 | 1477 | |||
| BC022142.1 | - | 9 | 759 | 5101205 | ||
| BC022602.1 | - | 7 | 628 | 4161204 | ||
| BC026651.1 | - | 13 | 1398 | 4987771 | ||
| BC027046.1 | - | 7 | 776 | 5362953 | ||
| BC029150.1 | - | 13 | 1470 | 4954376 | ||

Tmem214
Genomic Map