siRNA Details
| Gene Symbol: Foxk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080932.2 | NP_001074401.2 | 9 | 1924 | ||
| GenBank mRNA | AK150439.1 | - | 8 | 1368 | ||
| BC045204.1 | - | 3 | 488 | |||
| EU882160.1 | - | 9 | 1924 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxk2| Gene Symbol: Foxk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080932.2 | NP_001074401.2 | 9 | 1924 | ||
| GenBank mRNA | AK150439.1 | - | 8 | 1368 | ||
| BC045204.1 | - | 3 | 488 | |||
| EU882160.1 | - | 9 | 1924 | |||