siRNA Details
Gene Symbol: Foxk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080932.2 | NP_001074401.2 | 7 | 1667 | ||
GenBank mRNA | AK150439.1 | - | 7 | 1321 | ||
BC045204.1 | - | 2 | 441 | |||
EU882160.1 | - | 7 | 1667 |
