siRNA Details
| Gene Symbol: Rnf122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175136.2 | NP_780345.1 | 6 | 752 | ||
| GenBank mRNA | AK077856.1 | - | 6 | 775 | ||
| AK085376.1 | - | 6 | 734 | |||
| AK136206.1 | - | 6 | 773 | |||
| AK171823.1 | - | 6 | 775 | |||
| BC038399.1 | - | 6 | 752 | 4947596 | ||
| BC050268.1 | - | 5 | 442 | |||

Rnf122
Genomic Map