siRNA Details
Gene Symbol: Rnf122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175136.2 | NP_780345.1 | 1, 2 | 319 | ||
GenBank mRNA | AK077856.1 | - | 1, 2 | 339 | ||
AK085376.1 | - | 1, 2 | 301 | |||
AK136206.1 | - | 1, 2 | 337 | |||
AK162034.1 | - | 1, 2 | 337 | |||
AK171823.1 | - | 1, 2 | 339 | |||
BC038399.1 | - | 1, 2 | 319 | 4947596 |
