siRNA Details
| Gene Symbol: Klhdc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033039.2 | NP_001028211.1 | 2 | 676 | ||
| GenBank mRNA | AK076812.1 | - | 2 | 399 | ||
| AK140290.1 | - | 2 | 677 | |||
| BC141196.1 | - | 2 | 589 | 9055808 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhdc9| Gene Symbol: Klhdc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033039.2 | NP_001028211.1 | 2 | 676 | ||
| GenBank mRNA | AK076812.1 | - | 2 | 399 | ||
| AK140290.1 | - | 2 | 677 | |||
| BC141196.1 | - | 2 | 589 | 9055808 | ||