siRNA Details
| Gene Symbol: Prpf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133701.2 | NP_598462.1 | 8 | 1103 | ||
| GenBank mRNA | AK081998.1 | - | 8 | 1038 | ||
| AK145461.1 | - | 8 | 1106 | |||
| AK166890.1 | - | 8 | 1055 | |||
| AK178683.1 | - | 1 | 107 | |||
| AK189258.1 | - | 1 | 107 | |||
| AK219394.1 | - | 1 | 108 | |||
| BC014869.1 | - | 8 | 1030 | 3602158 | ||
| BC023691.1 | - | 8 | 1032 | 5344332 | ||
| BC025030.1 | - | 8 | 1049 | 4947416 | ||

Prpf6
Genomic Map