siRNA Details
| Gene Symbol: Cd177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026862.3 | NP_081138.2 | 15 | 1985 | ||
| GenBank mRNA | AK004472.1 | - | 2 | 145 | ||
| BC005615.1 | - | 10 | 1355 | |||
| BC027283.1 | - | 15 | 1985 | 2648499 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd177| Gene Symbol: Cd177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026862.3 | NP_081138.2 | 15 | 1985 | ||
| GenBank mRNA | AK004472.1 | - | 2 | 145 | ||
| BC005615.1 | - | 10 | 1355 | |||
| BC027283.1 | - | 15 | 1985 | 2648499 | ||